Assay Details
| Gene Symbol: LOC4331172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770713.2 | XP_015626199.1 | 2 - 3 | 314 | 66 | |
| GenBank mRNA | AK060869.1 | - | 2 - 3 | 298 | 66 | |
| AK104724.1 | - | 2 - 3 | 283 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015770713.2 transcripts.
Set Membership: |
|

LOC4331172
Genomic Map