Assay Details
| Gene Symbol: LOC4331172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770712.1 | XP_015626198.1 | 3 - 4 | 470 | 89 | |
| XM_015770713.2 | XP_015626199.1 | 3 - 4 | 449 | 89 | ||
| GenBank mRNA | AK060869.1 | - | 3 - 4 | 433 | 89 | |
| AK061365.1 | - | 3 - 4 | 445 | 89 | ||
| AK103008.1 | - | 3 - 4 | 476 | 89 | ||
| AK104724.1 | - | 3 - 4 | 418 | 89 | ||
More Information
Set Membership: |
|

LOC4331172
Genomic Map