Assay Details
| Gene Symbol: LOC4331262 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769136.2 | XP_015624622.1 | 5 - 6 | 632 | 91 | |
| GenBank mRNA | AK099439.1 | - | 4 - 5 | 676 | 91 | |
| AK099942.1 | - | 4 - 5 | 610 | 91 | ||
More Information
Set Membership: |
|

LOC4331262
Genomic Map