Assay Details
| Gene Symbol: LOC4331262 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769136.2 | XP_015624622.1 | 9 - 10 | 975 | 61 | |
| GenBank mRNA | AK099439.1 | - | 8 - 9 | 1019 | 61 | |
| AK099942.1 | - | 7 - 8 | 1059 | 61 | ||
More Information
Set Membership: |
|

LOC4331262
Genomic Map