Assay Details
| Gene Symbol: LOC4331382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773545.2 | XP_015629031.1 | 9 - 10 | 1564 | 98 | |
| XM_015773546.2 | XP_015629032.1 | 9 - 10 | 1564 | 98 | ||
| GenBank mRNA | AK063381.1 | - | 1 - 2 | 128 | 98 | |
More Information
Set Membership: |
|

LOC4331382
Genomic Map