Assay Details
| Gene Symbol: LOC4331445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775493.2 | XP_015630979.1 | 1 - 2 | 212 | 121 | |
| GenBank mRNA | AB003328.1 | - | 1 - 2 | 120 | 121 | |
| AK067641.1 | - | 1 - 2 | 140 | 121 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015775493.2 transcripts.
Set Membership: |
|

LOC4331445
Genomic Map