Assay Details
| Gene Symbol: LOC4331445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775493.2 | XP_015630979.1 | 3 - 4 | 514 | 117 | |
| GenBank mRNA | AB003328.1 | - | 3 - 4 | 422 | 117 | |
| AK067641.1 | - | 3 - 4 | 442 | 117 | ||
| AK104921.1 | - | 2 - 3 | 334 | 117 | ||
| AY332476.1 | - | 2 - 3 | 329 | 117 | ||
More Information
Set Membership: |
|

LOC4331445
Genomic Map