Assay Details
| Gene Symbol: LOC4331445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775493.2 | XP_015630979.1 | 6 - 7 | 723 | 67 | |
| GenBank mRNA | AB003328.1 | - | 6 - 7 | 631 | 67 | |
| AK067641.1 | - | 6 - 7 | 651 | 67 | ||
| AK104921.1 | - | 5 - 6 | 543 | 67 | ||
| AY332476.1 | - | 5 - 6 | 538 | 67 | ||
| AY551914.1 | - | 4 - 5 | 341 | 67 | ||
More Information
Set Membership: |
|

LOC4331445
Genomic Map