Assay Details
| Gene Symbol: LOC4331445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775493.2 | XP_015630979.1 | 7 - 8 | 761 | 72 | |
| GenBank mRNA | AB003328.1 | - | 7 - 8 | 669 | 72 | |
| AK067641.1 | - | 7 - 8 | 689 | 72 | ||
| AK104921.1 | - | 6 - 7 | 581 | 72 | ||
| AY332476.1 | - | 6 - 7 | 576 | 72 | ||
| AY551914.1 | - | 5 - 6 | 379 | 72 | ||
More Information
Set Membership: |
|

LOC4331445
Genomic Map