Assay Details
| Gene Symbol: LOC9271214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775313.2 | XP_015630799.1 | 3 | 1747 | 85 | |
| XM_015775314.2 | XP_015630800.1 | 3 | 1734 | 85 | ||
| GenBank mRNA | AK058297.1 | - | 1 | 1084 | 85 | |
More Information
Set Membership: |
|

LOC9271214
Genomic Map