Assay Details
Gene Symbol: LOC4332232 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015776326.2 | XP_015631812.1 | 1 - 2 | 230 | 73 | |
GenBank mRNA | AK062288.1 | - | 1 - 2 | 201 | 73 | |
AK287988.1 | - | 2133 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
