Assay Details
| Gene Symbol: LOC4332289 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776572.1 | XP_015632058.1 | 13 - 14 | 2381 | 75 | |
| GenBank mRNA | AK061989.1 | - | 6 - 7 | 779 | 75 | |
| AK067479.1 | - | 13 - 14 | 2220 | 75 | ||
More Information
Set Membership: |
|

LOC4332289
Genomic Map