Assay Details
| Gene Symbol: LOC4332289 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776572.1 | XP_015632058.1 | 9 - 10 | 1952 | 102 | |
| GenBank mRNA | AK061989.1 | - | 2 - 3 | 350 | 102 | |
| AK067479.1 | - | 9 - 10 | 1791 | 102 | ||
More Information
Set Membership: |
|

LOC4332289
Genomic Map