Assay Details
| Gene Symbol: LOC4332350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777396.2 | XP_015632882.1 | 2 - 3 | 170 | 73 | |
| XM_015777398.2 | XP_015632884.1 | 2 - 3 | 171 | 73 | ||
| GenBank mRNA | AK065578.1 | - | 2 - 3 | 108 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015777396.2, XM_015777398.2 transcripts.
Set Membership: |
|

LOC4332350
Genomic Map