Assay Details
| Gene Symbol: LOC4332468 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772625.2 | 3 - 4 | 459 | 91 | ||
| GenBank mRNA | AK110712.1 | - | 3 - 4 | 442 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4332468| Gene Symbol: LOC4332468 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772625.2 | 3 - 4 | 459 | 91 | ||
| GenBank mRNA | AK110712.1 | - | 3 - 4 | 442 | 91 | |
Set Membership: |
|