Assay Details
| Gene Symbol: LOC4332468 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772625.2 | 5 - 6 | 547 | 107 | ||
| GenBank mRNA | AK110712.1 | - | 5 - 6 | 530 | 107 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4332468| Gene Symbol: LOC4332468 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772625.2 | 5 - 6 | 547 | 107 | ||
| GenBank mRNA | AK110712.1 | - | 5 - 6 | 530 | 107 | |
Set Membership: |
|