Assay Details
| Gene Symbol: LOC4332486 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772964.2 | XP_015628450.1 | 3 - 4 | 431 | 99 | |
| GenBank mRNA | AK063663.1 | - | 3 - 4 | 411 | 99 | |
More Information
Set Membership: |
|

LOC4332486
Genomic Map