Assay Details
| Gene Symbol: LOC4333230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777785.2 | XP_015633271.1 | 2 - 3 | 431 | 92 | |
| GenBank mRNA | AK111867.1 | - | 2 - 3 | 378 | 92 | |
More Information
Set Membership: |
|

LOC4333230
Genomic Map