Assay Details
| Gene Symbol: LOC9272009 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774351.2 | XP_015629837.1 | 3 - 4 | 468 | 88 | |
| GenBank mRNA | AK063769.1 | - | 1665 | 88 | ||
| AK241140.1 | - | 3 - 4 | 475 | 88 | ||
| AK287904.1 | - | 3 - 4 | 425 | 88 | ||
More Information
Set Membership: |
|

LOC9272009
Genomic Map