Assay Details
| Gene Symbol: LOC4334405 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775565.2 | XP_015631051.1 | 3 - 4 | 391 | 88 | |
| XM_015775566.2 | XP_015631052.1 | 3 - 4 | 311 | 88 | ||
| GenBank mRNA | AB024962.1 | - | 2 - 3 | 231 | 88 | |
| AK071839.1 | - | 3 - 4 | 359 | 88 | ||
| AY332470.1 | - | 2 - 3 | 288 | 88 | ||
More Information
Set Membership: |
|

LOC4334405
Genomic Map