Assay Details
| Gene Symbol: LOC4334405 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775565.2 | XP_015631051.1 | 6 - 7 | 815 | 76 | |
| XM_015775566.2 | XP_015631052.1 | 6 - 7 | 735 | 76 | ||
| GenBank mRNA | AB024962.1 | - | 5 - 6 | 655 | 76 | |
| AK071839.1 | - | 6 - 7 | 782 | 76 | ||
| AY332470.1 | - | 5 - 6 | 712 | 76 | ||
More Information
Set Membership: |
|

LOC4334405
Genomic Map