Assay Details
| Gene Symbol: LOC4334582 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772883.2 | XP_015628369.1 | 9 - 10 | 1663 | 103 | |
| XM_015772884.2 | XP_015628370.1 | 9 - 10 | 1651 | 103 | ||
| GenBank mRNA | AY685113.1 | - | 9 - 10 | 1094 | 103 | |
More Information
Set Membership: |
|

LOC4334582
Genomic Map