Assay Details
| Gene Symbol: LOC4334610 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774473.2 | XP_015629959.1 | 1 | 578 | 123 | |
| GenBank mRNA | AK070075.1 | - | 1 | 550 | 123 | |
More Information
Set Membership: |
|

LOC4334610
Genomic Map