Assay Details
| Gene Symbol: LOC4334673 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776911.2 | XP_015632397.1 | 1 - 2 | 168 | 95 | |
| GenBank mRNA | AK103569.1 | - | 1 - 2 | 107 | 95 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015776911.2 transcripts.
Set Membership: |
|

LOC4334673
Genomic Map