Assay Details
| Gene Symbol: LOC4334690 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772726.2 | XP_015628212.1 | 2 - 3 | 206 | 105 | |
| GenBank mRNA | AK073655.1 | - | 2 - 3 | 184 | 105 | |
| AK103800.1 | - | 2 - 3 | 163 | 105 | ||
| AK104068.1 | - | 2 - 3 | 184 | 105 | ||
More Information
Set Membership: |
|

LOC4334690
Genomic Map