Assay Details
| Gene Symbol: LOC4334895 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780687.2 | XP_015636173.1 | 2 - 3 | 595 | 69 | |
| GenBank mRNA | AK060850.1 | - | 2 - 3 | 428 | 69 | |
| AK066032.1 | - | 2 - 3 | 466 | 69 | ||
More Information
Set Membership: |
|

LOC4334895
Genomic Map