Assay Details
| Gene Symbol: LOC4334935 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781294.2 | XP_015636780.1 | 1 - 2 | 3490 | 77 | |
| GenBank mRNA | AK067211.1 | - | 1 - 2 | 3456 | 77 | |
| AK121138.1 | - | 1 - 2 | 3449 | 77 | ||
More Information
Set Membership: |
|

LOC4334935
Genomic Map