Assay Details
| Gene Symbol: LOC4335202 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778949.2 | XP_015634435.1 | 7 - 8 | 630 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4335202| Gene Symbol: LOC4335202 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778949.2 | XP_015634435.1 | 7 - 8 | 630 | 103 | |
Set Membership: |
|