Assay Details
| Gene Symbol: LOC4335223 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778705.2 | XP_015634191.1 | 3 - 4 | 983 | 62 | |
| GenBank mRNA | AK071484.1 | - | 3 - 4 | 985 | 62 | |
| AK099270.1 | - | 3 - 4 | 982 | 62 | ||
| Gene Symbol: LOC9270335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK287874.1 | - | 1635 | 62 | ||
More Information
Set Membership: |
|

LOC4335223
Genomic Map