Assay Details
| Gene Symbol: LOC4335629 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778976.1 | XP_015634462.1 | 1 - 2 | 450 | 71 | |
| GenBank mRNA | AK067334.1 | - | 1 - 2 | 457 | 71 | |
| AK104271.1 | - | 1 - 2 | 360 | 71 | ||
| AK104769.1 | - | 1 - 2 | 360 | 71 | ||
More Information
Set Membership: |
|

LOC4335629
Genomic Map