Assay Details
| Gene Symbol: LOC4335701 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780045.2 | XP_015635531.1 | 1 - 2 | 134 | 59 | |
| GenBank mRNA | AK063198.1 | - | 1 - 2 | 112 | 59 | |
| BT131973.1 | - | 1 - 2 | 72 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015780045.2 transcripts.
Set Membership: |
|

LOC4335701
Genomic Map