Assay Details
| Gene Symbol: LOC4335701 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780045.2 | XP_015635531.1 | 2 - 3 | 228 | 72 | |
| GenBank mRNA | AK063198.1 | - | 2 - 3 | 206 | 72 | |
| BT131973.1 | - | 2 - 3 | 166 | 72 | ||
| D89726.1 | - | 1 - 2 | 99 | 72 | ||
More Information
Set Membership: |
|

LOC4335701
Genomic Map