Assay Details
| Gene Symbol: LOC4335767 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778059.1 | XP_015633545.1 | 358 | 79 | ||
| GenBank mRNA | AK065005.1 | - | 157 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015778059.1 transcripts.
Set Membership: |
|

LOC4335767
Genomic Map