Assay Details
| Gene Symbol: LOC4335903 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781487.2 | XP_015636973.1 | 4 - 5 | 1228 | 65 | |
| GenBank mRNA | AK100857.1 | - | 4 - 5 | 1030 | 65 | |
More Information
Set Membership: |
|

LOC4335903
Genomic Map