Assay Details
| Gene Symbol: LOC4335963 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778094.2 | XP_015633580.1 | 1 - 2 | 299 | 84 | |
| GenBank mRNA | AK099454.1 | - | 1 - 2 | 297 | 84 | |
More Information
Set Membership: |
|

LOC4335963
Genomic Map