Assay Details
| Gene Symbol: LOC4335968 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777910.2 | XP_015633396.1 | 3 - 4 | 495 | 121 | |
| GenBank mRNA | AK073743.1 | - | 3 - 4 | 500 | 121 | |
| AK103814.1 | - | 3 - 4 | 498 | 121 | ||
More Information
Set Membership: |
|

LOC4335968
Genomic Map