Assay Details
| Gene Symbol: LOC4335968 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777910.2 | XP_015633396.1 | 5 - 6 | 724 | 133 | |
| GenBank mRNA | AB118009.1 | - | 2 - 3 | 166 | 133 | |
| AK073743.1 | - | 5 - 6 | 729 | 133 | ||
| AK103814.1 | - | 5 - 6 | 727 | 133 | ||
More Information
Set Membership: |
|

LOC4335968
Genomic Map