Assay Details
| Gene Symbol: LOC4335983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778092.2 | XP_015633578.1 | 2 - 3 | 844 | 85 | |
| XM_015778093.2 | XP_015633579.1 | 2 - 3 | 844 | 85 | ||
| GenBank mRNA | AK069397.1 | - | 2 - 3 | 712 | 85 | |
| AK105021.2 | - | 2 - 3 | 849 | 85 | ||
More Information
Set Membership: |
|

LOC4335983
Genomic Map