Assay Details
| Gene Symbol: LOC4335983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778092.2 | XP_015633578.1 | 4 - 5 | 1108 | 93 | |
| XM_015778093.2 | XP_015633579.1 | 4 - 5 | 1108 | 93 | ||
| GenBank mRNA | AK069397.1 | - | 4 - 5 | 976 | 93 | |
| AK105021.2 | - | 4 - 5 | 1113 | 93 | ||
More Information
Set Membership: |
|

LOC4335983
Genomic Map