Assay Details
| Gene Symbol: LOC4335983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778093.2 | XP_015633579.1 | 6 - 7 | 1283 | 84 | |
| GenBank mRNA | AK069397.1 | - | 6 - 7 | 1151 | 84 | |
| AK105021.2 | - | 6 - 7 | 1288 | 84 | ||
More Information
Set Membership: |
|

LOC4335983
Genomic Map