Assay Details
| Gene Symbol: LOC4336015 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781757.2 | XP_015637243.1 | 1 - 2 | 367 | 67 | |
| GenBank mRNA | AK070018.1 | - | 1 - 2 | 213 | 67 | |
More Information
Set Membership: |
|

LOC4336015
Genomic Map