Assay Details
| Gene Symbol: LOC4336146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780551.2 | XP_015636037.1 | 6 - 7 | 3033 | 66 | |
| XM_015780553.2 | XP_015636039.1 | 6 - 7 | 3027 | 66 | ||
| XM_015780557.2 | XP_015636043.1 | 6 - 7 | 3016 | 66 | ||
| GenBank mRNA | AK070962.1 | - | 3 - 4 | 306 | 66 | |
More Information
Set Membership: |
|

LOC4336146
Genomic Map