Assay Details
| Gene Symbol: LOC4336146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780551.2 | XP_015636037.1 | 12 - 13 | 3857 | 101 | |
| XM_015780553.2 | XP_015636039.1 | 12 - 13 | 3851 | 101 | ||
| XM_015780557.2 | XP_015636043.1 | 12 - 13 | 3840 | 101 | ||
| GenBank mRNA | AK070962.1 | - | 9 - 10 | 1130 | 101 | |
More Information
Set Membership: |
|

LOC4336146
Genomic Map