Assay Details
| Gene Symbol: LOC4336455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779014.2 | XP_015634500.1 | 1 | 777 | 82 | |
| GenBank mRNA | AK106723.1 | - | 1 | 763 | 82 | |
| AY166458.1 | - | 1 | 751 | 82 | ||
More Information
Set Membership: |
|

LOC4336455
Genomic Map