Assay Details
Gene Symbol: LOC4336481 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780364.2 | XP_015635850.1 | 1 - 2 | 730 | 66 | |
GenBank mRNA | AK066705.1 | - | 1 - 2 | 701 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
