Assay Details
Gene Symbol: LOC4336481 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780364.2 | XP_015635850.1 | 7 - 8 | 1135 | 83 | |
GenBank mRNA | AK066705.1 | - | 7 - 8 | 1106 | 83 | |
BT129736.1 | - | 4 - 5 | 281 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
