Assay Details
| Gene Symbol: LOC4336627 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780454.2 | XP_015635940.1 | 5 - 6 | 708 | 67 | |
| GenBank mRNA | AK058931.1 | - | 3 - 4 | 317 | 67 | |
| AK062772.1 | - | 5 - 6 | 596 | 67 | ||
| BT129160.1 | - | 5 - 6 | 589 | 67 | ||
| BT129653.1 | - | 1 - 2 | 51 | 67 | ||
More Information
Set Membership: |
|

LOC4336627
Genomic Map