Assay Details
| Gene Symbol: LOC4336765 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778633.2 | XP_015634119.1 | 2 - 3 | 579 | 63 | |
| GenBank mRNA | AK100853.1 | - | 2 - 3 | 569 | 63 | |
More Information
Set Membership: |
|

LOC4336765
Genomic Map