Assay Details
| Gene Symbol: LOC4336971 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780125.2 | XP_015635611.1 | 2 - 3 | 1188 | 94 | |
| GenBank mRNA | AK069804.1 | - | 2 - 3 | 1013 | 94 | |
| AY323915.1 | - | 2 - 3 | 839 | 94 | ||
More Information
Set Membership: |
|

LOC4336971
Genomic Map