Assay Details
| Gene Symbol: LOC9267450 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780472.2 | XP_015635958.1 | 3 - 4 | 754 | 67 | |
| GenBank mRNA | AK068148.1 | - | 4 - 5 | 2941 | 67 | |
More Information
Set Membership: |
|

LOC9267450
Genomic Map